Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000748.2 | NP_000739.1 | 5 | 656 | ||
GenBank mRNA | AK313470.1 | - | 5 | 425 | ||
BC075040.2 | - | 5 | 421 | 30915357 | ||
BC075041.2 | - | 5 | 421 | 30915573 | ||
U62437.1 | - | 5 | 656 | |||
X53179.1 | - | 5 | 558 | |||
Y08415.1 | - | 5 | 392 |