Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT1 SIRNA Assays ›
Gene Symbol:
RHOT1
Gene Name:
ras homolog family member T1
Gene Aliases:
ARHT1, MIRO-1, MIRO1
Chromosome Location:
Chr.17: 32142454 - 32253374 on Build GRCh38
UniGene ID:
Hs.655325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30649
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033566.2
NP_001028738.1
14
1393
NM_001033567.2
NP_001028739.2
15
1517
NM_001033568.2
NP_001028740.1
14
1393
NM_001288754.1
NP_001275683.1
14
1393
NM_001288755.1
NP_001275684.1
14
1389
NM_001288758.1
NP_001275687.1
15
1517
NM_018307.4
NP_060777.3
14
1393
NR_110083.1
14
1393
XM_011524969.2
XP_011523271.1
13
1117
XM_011524971.2
XP_011523273.2
14
1189
XM_011524973.2
XP_011523275.2
14
1189
XM_011524975.2
XP_011523277.1
15
1646
XM_011524976.2
XP_011523278.2
14
1189
XM_017024819.1
XP_016880308.1
15
1421
XM_017024820.1
XP_016880309.1
13
1117
XM_017024821.1
XP_016880310.1
14
1189
XM_017024822.1
XP_016880311.1
14
1418
XM_017024823.1
XP_016880312.1
14
1189
XM_017024824.1
XP_016880313.1
15
1524
XM_017024825.1
XP_016880314.1
15
1616
GenBank mRNA
AJ496730.1
-
14
1381
AJ517412.1
-
14
1154
AK001902.1
-
8
757
AK022695.1
-
7
687
AK294407.1
-
14
1167
AL136929.1
-
14
1217
AY094972.1
-
14
1270
BC041114.1
-
14
1327
BC051818.1
-
13
1104
BC060781.1
-
14
1393
4823115
BC068463.1
-
14
1281
BC125104.1
-
14
1222
40118340
BC125105.1
-
14
1222
40118341
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00430266_m1
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
molecular_function
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0