Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292054.1 | NP_001278983.1 | 6 | 716 | ||
NM_018317.3 | NP_060787.2 | 9 | 911 | |||
XM_011513852.2 | XP_011512154.1 | 9 | 751 | |||
XM_017008363.1 | XP_016863852.1 | 9 | 751 | |||
XM_017008364.1 | XP_016863853.1 | 9 | 751 | |||
GenBank mRNA | AB449905.1 | - | 6 | 446 | ||
AK001944.1 | - | 9 | 751 | |||
AK225395.1 | - | 9 | 751 | |||
AK289631.1 | - | 9 | 751 | |||
AK310795.1 | - | 9 | 758 | |||
BC031642.1 | - | 9 | 707 | 5175317 |