Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018361.3 | NP_060831.2 | 7, 8 | 1181 | ||
XM_011534751.2 | XP_011533053.1 | 5, 6 | 701 | |||
GenBank mRNA | AF375789.1 | - | 7, 8 | 1143 | ||
AK002072.1 | - | 7, 8 | 845 | |||
AK310545.1 | - | 2, 3 | 320 | |||
AL136587.1 | - | 7, 8 | 871 | |||
BC023550.2 | - | 7, 8 | 1125 | 4129973 | ||
BC068519.1 | - | 7, 8 | 845 | |||
BC080537.1 | - | 7, 8 | 1150 | 5015128 |