Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308277.1 | NP_001295206.1 | 4 | 517 | ||
NM_018381.3 | NP_060851.2 | 4 | 517 | |||
XM_011528121.1 | XP_011526423.1 | 6 | 687 | |||
XM_017026934.1 | XP_016882423.1 | 4 | 455 | |||
GenBank mRNA | AK001098.1 | - | 223 | |||
AK002148.1 | - | 4 | 303 | |||
AK096142.1 | - | 4 | 532 | |||
AL137694.1 | - | 4 | 219 | |||
BC035817.2 | - | 4 | 309 | 5554801 |