Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018385.2 | NP_060855.2 | 4, 5 | 756 | ||
GenBank mRNA | AK002163.1 | - | 4, 5 | 489 | ||
AK225094.1 | - | 4, 5 | 457 | |||
AL136897.1 | - | 4, 5 | 461 | |||
BC015042.1 | - | 4, 5 | 461 | |||
BC040119.1 | - | 4, 5 | 441 | |||
BC058891.1 | - | 4, 5 | 447 | |||
BC068500.1 | - | 4, 5 | 470 | 30343878 |