Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 11 | 1415 | ||
NM_018401.2 | NP_060871.1 | 10 | 1305 | |||
XM_005247982.3 | XP_005248039.1 | 5 | 1812 | |||
XM_005247983.4 | XP_005248040.1 | 5 | 1070 | |||
XM_011513497.1 | XP_011511799.1 | 9 | 1409 | |||
XM_011513501.2 | XP_011511803.1 | 6 | 604 | |||
XM_011513504.1 | XP_011511806.1 | 5 | 458 | |||
XM_017008377.1 | XP_016863866.1 | 5 | 1857 | |||
GenBank mRNA | AJ250839.1 | - | 10 | 1022 | ||
AK292696.1 | - | 10 | 1272 | |||
AK313846.1 | - | 10 | 1307 | |||
AL832436.1 | - | 4 | 358 | |||
AY358353.1 | - | 10 | 1049 | |||
BC038238.1 | - | 10 | 999 | 5744969 | ||
BC069203.1 | - | 11 | 1336 |