Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018518.4 | NP_060988.3 | 13 | 1811 | ||
NM_182751.2 | NP_877428.1 | 13 | 1814 | |||
XM_011519538.2 | XP_011517840.1 | 13 | 1784 | |||
GenBank mRNA | AB042719.1 | - | 13 | 1778 | ||
AF119869.1 | - | 2 | 150 | |||
AK055695.1 | - | 13 | 1787 | |||
AK292701.1 | - | 13 | 1789 | |||
AL136840.1 | - | 12 | 1777 | |||
BC004876.2 | - | 6 | 729 | |||
BC009108.1 | - | 8 | 1029 | |||
BC101727.1 | - | 13 | 1783 | 8069233 | ||
BC143490.1 | - | 13 | 1776 | 9051998 |