Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018518.4 | NP_060988.3 | 3 | 289 | ||
NM_182751.2 | NP_877428.1 | 3 | 289 | |||
XM_011519538.2 | XP_011517840.1 | 3 | 259 | |||
GenBank mRNA | AB042719.1 | - | 3 | 256 | ||
AK055695.1 | - | 3 | 262 | |||
AK292701.1 | - | 3 | 264 | |||
AL136840.1 | - | 2 | 255 | |||
BC101727.1 | - | 3 | 258 | 8069233 | ||
BC143490.1 | - | 3 | 254 | 9051998 | ||
DA024424.1 | - | 3 | 289 |