Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 8 | 1240 | ||
GenBank mRNA | AF304463.1 | - | 8 | 1011 | ||
AF365927.1 | - | 8 | 1227 | |||
AF365928.1 | - | 4 | 654 | |||
AJ243500.2 | - | 8 | 1211 | |||
AJ243501.2 | - | 8 | 1211 | |||
AJ487964.2 | - | 9 | 1180 | |||
AK075103.1 | - | 3 | 382 | |||
AK291707.1 | - | 8 | 1241 | |||
AY203947.1 | - | 5 | 1421 | |||
KF534785.1 | - | 8 | 1240 |