Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 2 | 415 | ||
GenBank mRNA | AF304463.1 | - | 2 | 186 | ||
AF365927.1 | - | 2 | 402 | |||
AJ243500.2 | - | 2 | 386 | |||
AJ243501.2 | - | 2 | 386 | |||
AJ487964.2 | - | 3 | 355 | |||
AK291707.1 | - | 2 | 416 | |||
AY203947.1 | - | 1 | 70 | |||
KF534785.1 | - | 2 | 415 |