Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 15 | 2144 | ||
GenBank mRNA | AF304463.1 | - | 15 | 1915 | ||
AF365927.1 | - | 15 | 2131 | |||
AF365928.1 | - | 11 | 1558 | |||
AJ243500.2 | - | 15 | 2115 | |||
AJ243501.2 | - | 15 | 2115 | |||
AJ487964.2 | - | 16 | 2084 | |||
AK075103.1 | - | 9 | 1645 | |||
AK075206.1 | - | 5 | 514 | |||
AK291707.1 | - | 15 | 2145 | |||
AY203947.1 | - | 12 | 2325 | |||
KF534785.1 | - | 15 | 2144 |