Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018666.2 | NP_061136.2 | 16 | 2150 | ||
XM_017029621.1 | XP_016885110.1 | 16 | 2158 | |||
XM_017029622.1 | XP_016885111.1 | 16 | 2069 | |||
XM_017029623.1 | XP_016885112.1 | 16 | 2032 | |||
XM_017029624.1 | XP_016885113.1 | 15 | 1972 | |||
XM_017029625.1 | XP_016885114.1 | 16 | 2065 | |||
XM_017029626.1 | XP_016885115.1 | 10 | 1274 | |||
XM_017029627.1 | XP_016885116.1 | 6 | 1065 | |||
GenBank mRNA | AJ278111.1 | - | 16 | 2150 | ||
AK310412.1 | - | 15 | 2106 | |||
BC144260.1 | - | 9 | 903 | 9052780 |