Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF27 SIRNA Assays ›
Gene Symbol:
KIF27
Gene Name:
kinesin family member 27
Gene Aliases:
-
Chromosome Location:
Chr.9: 83832795 - 83921539 on Build GRCh38
UniGene ID:
Hs.697514
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31015
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271927.1
NP_001258856.1
14, 15
3349
NM_001271928.1
NP_001258857.1
13, 14
3256
NM_017576.2
NP_060046.1
15, 16
3547
XM_011518848.2
XP_011517150.1
16, 17
3626
XM_011518849.2
XP_011517151.1
17, 18
3742
XM_011518850.2
XP_011517152.1
16, 17
3715
XM_011518854.2
XP_011517156.1
15, 16
3452
XM_011518857.2
XP_011517159.1
17, 18
3311
XM_011518858.2
XP_011517160.1
13, 14
2099
XM_017014900.1
XP_016870389.1
18, 19
3894
XM_017014901.1
XP_016870390.1
18, 19
3776
XM_017014902.1
XP_016870391.1
17, 18
3822
XM_017014903.1
XP_016870392.1
16, 17
3767
XM_017014904.1
XP_016870393.1
16, 17
3664
XM_017014905.1
XP_016870394.1
16, 17
3647
XM_017014906.1
XP_016870395.1
16, 17
3629
XM_017014907.1
XP_016870396.1
15, 16
3569
XM_017014908.1
XP_016870397.1
14, 15
3476
XM_017014909.1
XP_016870398.1
16, 17
3447
XM_017014910.1
XP_016870399.1
14, 15
3271
XM_017014911.1
XP_016870400.1
14, 15
2941
XM_017014912.1
XP_016870401.1
14, 15
2586
XM_017014913.1
XP_016870402.1
15, 16
2778
XM_017014915.1
XP_016870404.1
14, 15
2532
XM_017014916.1
XP_016870405.1
16, 17
3353
GenBank mRNA
AL133654.1
-
10, 11
1662
AY237536.1
-
15, 16
3509
AY237537.1
-
14, 15
3311
AY237538.1
-
13, 14
3218
BC140788.1
-
15, 16
3508
9021656
HM370398.1
-
7, 8
1007
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cilium movement
microtubule-based movement
ventricular system development
cilium assembly
microtubule motor activity
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0