Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017797.3 | NP_060267.2 | 8 | 1348 | ||
XM_005259593.1 | XP_005259650.1 | 8 | 1113 | |||
XM_011528127.1 | XP_011526429.1 | 6 | 714 | |||
GenBank mRNA | AF355797.1 | - | 8 | 1348 | ||
AK000393.1 | - | 5 | 1142 | |||
AK027481.1 | - | 6 | 710 | |||
AK056818.1 | - | 2 | 1483 | |||
AK225831.1 | - | 8 | 1347 | |||
AL360275.1 | - | 5 | 579 | |||
AY007116.1 | - | 8 | 1070 | |||
BC000564.2 | - | 5 | 638 | |||
BC008035.2 | - | 5 | 632 | |||
BC053986.1 | - | 4 | 533 | |||
BC069199.1 | - | 6 | 774 | |||
BC094820.1 | - | 8 | 1082 | |||
CR457230.1 | - | 4 | 385 |