Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001828.5 | NP_001819.2 | 2, 3 | 171 | ||
GenBank mRNA | AV715261.1 | - | 2, 3 | 170 | ||
AV762486.1 | - | 2, 3 | 168 | |||
BC119711.1 | - | 2, 3 | 122 | 40117193 | ||
BC119712.1 | - | 2, 3 | 122 | 40117194 | ||
FJ613334.1 | - | 2, 3 | 114 | |||
FJ613335.1 | - | 2, 3 | 123 | |||
L01664.1 | - | 2, 3 | 127 |