Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 14 | 2102 | ||
NM_017988.5 | NP_060458.3 | 14 | 2254 | |||
XM_017019618.1 | XP_016875107.1 | 15 | 2114 | |||
XM_017019619.1 | XP_016875108.1 | 15 | 2266 | |||
XM_017019620.1 | XP_016875109.1 | 16 | 2253 | |||
GenBank mRNA | AB037781.1 | - | 12 | 1420 | ||
AK001597.1 | - | 4 | 414 | |||
AK024274.1 | - | 10 | 1263 | |||
AK027551.1 | - | 10 | 1320 | |||
AK125091.1 | - | 14 | 2239 | |||
AK292980.1 | - | 14 | 2028 | |||
BC012387.2 | - | 5 | 547 | |||
BC063798.1 | - | 14 | 1867 | 6473586 |