Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31244
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
6
823
NR_047653.1
6
884
NR_047654.1
6
884
NR_047655.1
5
732
NR_047656.1
5
732
NR_047657.1
6
823
NR_047658.1
6
800
XM_005263982.2
XP_005264039.1
6
822
XM_005263984.1
XP_005264041.1
6
822
XM_005263987.1
XP_005264044.1
6
822
XM_011511454.2
XP_011509756.1
6
822
XM_011511455.2
XP_011509757.1
6
791
XM_011511456.2
XP_011509758.1
6
825
XM_011511457.2
XP_011509759.1
6
744
XM_011511458.2
XP_011509760.1
6
822
XM_011511459.2
XP_011509761.1
6
822
XM_011511460.2
XP_011509762.1
6
822
XM_011511461.2
XP_011509763.1
6
822
XM_011511462.2
XP_011509764.1
5
651
XM_011511463.2
XP_011509765.1
6
835
XM_011511464.2
XP_011509766.1
6
894
XM_011511466.2
XP_011509768.1
6
835
XM_011511467.2
XP_011509769.1
5
677
XM_017004470.1
XP_016859959.1
6
822
XM_017004471.1
XP_016859960.1
5
729
XM_017004472.1
XP_016859961.1
6
835
XM_017004473.1
XP_016859962.1
6
835
XM_017004474.1
XP_016859963.1
6
730
GenBank mRNA
AB037731.1
-
6
775
AF311326.1
-
8
636
AK000943.1
-
4
724
AK023813.1
-
6
851
AK302646.1
-
5
673
AK304011.1
-
5
732
AK311617.1
-
5
665
AK314367.1
-
6
760
AY050169.1
-
6
851
BC015469.2
-
7
817
BC024276.1
-
6
760
BC032746.1
-
6
894
BC051763.1
-
4
454
BC063792.1
-
6
753
6504387
CB123459.1
-
4
356
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0