Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168347.2 | NP_001161819.1 | 4 | 525 | ||
NM_001168348.2 | NP_001161820.1 | 4 | 411 | |||
NM_018044.4 | NP_060514.1 | 4 | 525 | |||
NM_148956.3 | NP_683759.1 | 4 | 525 | |||
XM_005277604.4 | XP_005277661.1 | 4 | 502 | |||
GenBank mRNA | AF412028.1 | - | 4 | 464 | ||
AF420249.1 | - | 4 | 456 | |||
AK001129.1 | - | 4 | 464 | |||
AK098619.1 | - | 4 | 484 | |||
AK125667.1 | - | 3 | 1147 | |||
AK126375.1 | - | 4 | 348 | |||
AK298221.1 | - | 4 | 474 | |||
AK315361.1 | - | 4 | 466 | |||
BC008084.2 | - | 4 | 465 | 2823604 | ||
CB154882.1 | - | 4 | 525 |