Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300750.1 | NP_001287679.1 | 14, 15 | 1627 | ||
NM_001300751.1 | NP_001287680.1 | 13, 14 | 1365 | |||
NM_018113.3 | NP_060583.2 | 13, 14 | 1425 | |||
XM_011538563.2 | XP_011536865.1 | 13, 14 | 1579 | |||
XM_011538564.2 | XP_011536866.1 | 13, 14 | 1952 | |||
XM_011538565.2 | XP_011536867.1 | 12, 13 | 2201 | |||
XM_011538566.2 | XP_011536868.1 | 12, 13 | 1175 | |||
XM_017019626.1 | XP_016875115.1 | 13, 14 | 1827 | |||
XM_017019627.1 | XP_016875116.1 | 14, 15 | 1556 | |||
XM_017019628.1 | XP_016875117.1 | 13, 14 | 1414 | |||
XM_017019629.1 | XP_016875118.1 | 14, 15 | 2411 | |||
XM_017019630.1 | XP_016875119.1 | 10, 11 | 3387 | |||
XM_017019631.1 | XP_016875120.1 | 12, 13 | 1115 | |||
GenBank mRNA | AF260728.1 | - | 13, 14 | 1384 | ||
AK001356.1 | - | 14, 15 | 1408 | |||
AK001651.1 | - | 13, 14 | 1315 | |||
AL137599.1 | - | 10, 11 | 818 | |||
AY358572.1 | - | 13, 14 | 1389 | |||
BC008337.1 | - | 13, 14 | 1295 | 3506347 | ||
BC015015.1 | - | 13, 14 | 1154 | 3910520 | ||
BC031550.1 | - | 13, 14 | 1186 | 5194023 | ||
KU178711.1 | - | 13, 14 | 1080 | |||
KU178712.1 | - | 12, 13 | 1063 | |||
KU178713.1 | - | 12, 13 | 1046 | |||
KU178714.1 | - | 13, 14 | 1020 |