Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136123.1 | NP_001129595.1 | 5 | 2140 | ||
NM_018121.3 | NP_060591.3 | 5 | 2140 | |||
XM_005269965.2 | XP_005270022.1 | 5 | 1755 | |||
XM_011539944.2 | XP_011538246.1 | 5 | 4730 | |||
GenBank mRNA | AF460991.1 | - | 5 | 2140 | ||
AK057741.1 | - | 3 | 705 | |||
AK292565.1 | - | 5 | 1757 | |||
AL832343.1 | - | 5 | 1746 | |||
AL832383.1 | - | 5 | 1708 | |||
AL833126.1 | - | 5 | 1747 | |||
BC030565.1 | - | 5 | 1730 |