Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256944.1 | NP_001243873.1 | 3 | 545 | ||
NM_001830.3 | NP_001821.2 | 5 | 800 | |||
GenBank mRNA | AB019432.1 | - | 4 | 645 | ||
AF170492.1 | - | 3 | 370 | |||
AK225287.1 | - | 5 | 761 | |||
AK289564.1 | - | 5 | 747 | |||
AK299611.1 | - | 2 | 178 | |||
BC036068.1 | - | 4 | 420 | 5311366 | ||
BC130278.1 | - | 4 | 659 | 40146309 | ||
X77197.1 | - | 5 | 753 |