Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018238.3 | NP_060708.1 | 15 | 1291 | ||
XM_011516397.2 | XP_011514699.1 | 15 | 1757 | |||
GenBank mRNA | AJ278150.1 | - | 14 | 1147 | ||
AK001704.1 | - | 15 | 1159 | |||
AK222769.1 | - | 15 | 1153 | |||
AK304502.1 | - | 16 | 1178 | |||
BC022777.1 | - | 14 | 1145 | 3913448 |