Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31457
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
19
2622
NM_001280545.1
NP_001267474.1
20
2797
NM_001280546.1
NP_001267475.1
17
2390
NM_018262.3
NP_060732.2
18
2469
NM_052985.3
NP_443711.2
20
2799
NM_052989.2
NP_443715.1
19
2646
NM_052990.2
NP_443716.1
16
2313
XM_005247609.1
XP_005247666.1
18
2434
XM_006713689.1
XP_006713752.1
19
2611
XM_006713691.2
XP_006713754.1
18
2557
XM_006713692.2
XP_006713755.1
17
2380
XM_006713695.2
XP_006713758.1
19
2611
XM_011512972.2
XP_011511274.1
19
2611
XM_017006817.1
XP_016862306.1
19
2611
XM_017006818.1
XP_016862307.1
19
2611
XM_017006819.1
XP_016862308.1
18
2557
XM_017006820.1
XP_016862309.1
18
2557
XM_017006821.1
XP_016862310.1
18
2557
XM_017006822.1
XP_016862311.1
17
2402
XM_017006823.1
XP_016862312.1
18
2434
XM_017006824.1
XP_016862313.1
18
2434
XM_017006825.1
XP_016862314.1
17
2380
XM_017006826.1
XP_016862315.1
17
2380
XM_017006827.1
XP_016862316.1
17
2380
XM_017006828.1
XP_016862317.1
16
2225
XM_017006829.1
XP_016862318.1
16
2225
XM_017006830.1
XP_016862319.1
20
2609
XM_017006831.1
XP_016862320.1
18
2530
XM_017006832.1
XP_016862321.1
19
2509
XM_017006833.1
XP_016862322.1
15
2171
XM_017006834.1
XP_016862323.1
15
2171
XM_017006835.1
XP_016862324.1
17
2353
XM_017006836.1
XP_016862325.1
18
2378
XM_017006837.1
XP_016862326.1
10
1555
XM_017006838.1
XP_016862327.1
15
1995
GenBank mRNA
AF244930.1
-
19
2543
AF244931.1
-
20
2696
AF302154.1
-
19
2428
AK001759.1
-
18
2282
AK001955.1
-
8
1091
AK022525.1
-
18
2380
AK023000.1
-
18
2111
AK024435.1
-
17
2157
AK094922.1
-
3
512
AK096891.1
-
4
760
AK122965.1
-
12
2147
AK124140.1
-
20
2600
AK223095.1
-
18
2281
AK293852.1
-
15
2073
AK298526.1
-
15
1890
AK299903.1
-
17
2103
AK307463.1
-
6
876
AK315103.1
-
19
2446
AL133565.1
-
17
2223
AL833851.1
-
8
1093
BC003045.1
-
16
2105
2823536
BC004238.2
-
16
2105
2823536
BC028353.2
-
19
2386
4836844
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0