Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286.3 | NP_001277.1 | 3, 4 | 373 | ||
NR_046428.1 | 3, 4 | 373 | ||||
GenBank mRNA | AK289999.1 | - | 3, 4 | 278 | ||
BC117420.1 | - | 3, 4 | 230 | 40125971 | ||
BC117424.1 | - | 3, 4 | 230 | 40125975 | ||
D28475.2 | - | 3, 4 | 258 | |||
DA229615.1 | - | 3, 4 | 373 | |||
DA935587.1 | - | 3, 4 | 319 | |||
X83378.1 | - | 3, 4 | 232 | |||
X96391.1 | - | 3, 4 | 206 | |||
X99473.1 | - | 3, 4 | 206 | |||
X99474.1 | - | 3, 4 | 206 | |||
X99475.1 | - | 3, 4 | 206 |