Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018328.4 | NP_060798.2 | 12 | 4430 | ||
XM_011511470.2 | XP_011509772.1 | 12 | 5361 | |||
XM_011511472.2 | XP_011509774.1 | 10 | 4941 | |||
XM_017004482.1 | XP_016859971.1 | 8 | 4811 | |||
XM_017004483.1 | XP_016859972.1 | 8 | 4811 | |||
XM_017004484.1 | XP_016859973.1 | 8 | 4811 | |||
XM_017004485.1 | XP_016859974.1 | 8 | 4112 | |||
GenBank mRNA | AB040894.1 | - | 7 | 3723 | ||
AK055079.1 | - | 2 | 1216 | |||
AK090496.1 | - | 3 | 1319 | |||
BC150264.1 | - | 7 | 3723 | 9007212 | ||
EF542797.1 | - | 12 | 4421 |