Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291310.1 | NP_001278239.1 | 2281 | |||
NM_018461.4 | NP_060931.2 | 1244 | ||||
NR_033191.2 | 1230 | |||||
XM_006717914.3 | XP_006717977.1 | 7 | 954 | |||
XM_006717915.2 | XP_006717978.1 | 660 | ||||
XM_017016406.1 | XP_016871895.1 | 1306 | ||||
XM_017016407.1 | XP_016871896.1 | 5 | 1306 | |||
XM_017016408.1 | XP_016871897.1 | 512 | ||||
GenBank mRNA | AB040974.1 | - | 1719 | |||
AF220046.1 | - | 3 | 175 | |||
AK000402.1 | - | 4 | 438 | |||
AK293065.1 | - | 7 | 760 | |||
BC047379.1 | - | 7 | 1244 | 5267510 | ||
BC058076.1 | - | 2 | 238 | |||
BC072402.1 | - | 4 | 560 | 6048730 |