Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018704.2 | NP_061174.1 | 6 | 2098 | ||
XM_011541781.2 | XP_011540083.1 | 6 | 2169 | |||
XM_017001806.1 | XP_016857295.1 | 6 | 2536 | |||
GenBank mRNA | AK022544.1 | - | 5 | 2019 | ||
AL359565.1 | - | 1 | 547 | |||
BC016029.1 | - | 6 | 2038 | 3921313 | ||
DA238082.1 | - | 4 | 558 |