Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018704.2 | NP_061174.1 | 4 | 344 | ||
XM_011541781.2 | XP_011540083.1 | 4 | 415 | |||
XM_017001806.1 | XP_016857295.1 | 4 | 782 | |||
GenBank mRNA | AL598321.1 | - | 4 | 300 | ||
BC016029.1 | - | 4 | 284 | 3921313 | ||
DB236769.1 | - | 1 | 14 |