Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019117.4 | NP_061990.2 | 1 | 304 | ||
NM_057162.2 | NP_476503.1 | 1 | 304 | |||
GenBank mRNA | AB051474.1 | - | 1 | 250 | ||
AF284765.1 | - | 1 | 211 | |||
AF284766.1 | - | 1 | 211 | |||
AK056457.1 | - | 1 | 272 | |||
AL832514.1 | - | 1 | 244 | |||
BC140839.1 | - | 1 | 173 | 9021707 | ||
BC140840.1 | - | 1 | 173 | 9021708 | ||
BC142652.1 | - | 1 | 150 | 40148044 | ||
BC146677.1 | - | 1 | 150 | 40148894 | ||
BC161675.1 | - | 1 | 250 | 8862604 | ||
DB083610.1 | - | 1 | 304 |