Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146068.1 | NP_001139540.1 | 9 | 1056 | ||
NM_001748.4 | NP_001739.2 | 9 | 1278 | |||
XM_011544286.2 | XP_011542588.1 | 9 | 1455 | |||
GenBank mRNA | AB209855.1 | - | 9 | 1143 | ||
AF261089.1 | - | 9 | 1181 | |||
AK297800.1 | - | 9 | 1056 | |||
AK303952.1 | - | 5 | 652 | |||
AK315179.1 | - | 9 | 1180 | |||
AK316211.1 | - | 9 | 1099 | |||
BC007686.2 | - | 9 | 1278 | |||
BC011828.2 | - | 9 | 1278 | |||
BC021303.2 | - | 9 | 1172 | 5090865 | ||
M23254.1 | - | 9 | 1184 |