Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021047.2 | NP_066385.2 | 3 | 285 | ||
GenBank mRNA | AF038951.1 | - | 2 | 193 | ||
AK074757.1 | - | 3 | 277 | |||
BC014148.2 | - | 3 | 285 | |||
BC125063.1 | - | 3 | 264 | 40117279 |