Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001290.1 | NP_001001290.1 | 9 | 1131 | ||
NM_020041.2 | NP_064425.2 | 8 | 1120 | |||
XM_006713968.3 | XP_006714031.1 | 8 | 1232 | |||
XM_006713969.2 | XP_006714032.1 | 9 | 1093 | |||
XM_011513856.2 | XP_011512158.1 | 8 | 1257 | |||
XM_011513857.1 | XP_011512159.1 | 9 | 1093 | |||
XM_011513858.1 | XP_011512160.1 | 9 | 1295 | |||
XM_011513859.2 | XP_011512161.1 | 8 | 1227 | |||
XM_011513860.2 | XP_011512162.1 | 8 | 1227 | |||
XM_011513861.2 | XP_011512163.1 | 8 | 1229 | |||
XM_011513862.2 | XP_011512164.1 | 7 | 1104 | |||
XM_011513863.2 | XP_011512165.1 | 7 | 2783 | |||
XM_011513864.2 | XP_011512166.1 | 6 | 672 | |||
XM_011513865.2 | XP_011512167.1 | 8 | 1231 | |||
XM_011513866.2 | XP_011512168.1 | 8 | 1229 | |||
XM_011513867.2 | XP_011512169.1 | 5 | 1186 | |||
XM_011513868.2 | XP_011512170.1 | 8 | 1248 | |||
XM_017008457.1 | XP_016863946.1 | 8 | 1245 | |||
XM_017008458.1 | XP_016863947.1 | 8 | 1230 | |||
XM_017008459.1 | XP_016863948.1 | 6 | 764 | |||
XM_017008460.1 | XP_016863949.1 | 7 | 2777 | |||
GenBank mRNA | AK310895.1 | - | 9 | 1093 |