Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 37 | 4411 | ||
NR_027671.2 | 37 | 4564 | ||||
XM_006712634.3 | XP_006712697.1 | 37 | 4385 | |||
XM_006712635.3 | XP_006712698.1 | 36 | 4355 | |||
XM_006712636.3 | XP_006712699.1 | 37 | 4374 | |||
XM_017004507.1 | XP_016859996.1 | 38 | 4470 | |||
XM_017004508.1 | XP_016859997.1 | 38 | 4425 | |||
XM_017004509.1 | XP_016859998.1 | 23 | 2716 | |||
XM_017004510.1 | XP_016859999.1 | 19 | 3095 | |||
GenBank mRNA | AF227905.1 | - | 37 | 4271 | ||
AK074251.1 | - | 21 | 2435 | |||
AK074376.1 | - | 11 | 1406 | |||
AK293066.1 | - | 37 | 4564 | |||
BC041098.1 | - | 37 | 4561 | 5492334 |