Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 33 | 3822 | ||
NR_027671.2 | 33 | 3975 | ||||
XM_006712634.3 | XP_006712697.1 | 33 | 3796 | |||
XM_006712635.3 | XP_006712698.1 | 32 | 3766 | |||
XM_006712636.3 | XP_006712699.1 | 33 | 3785 | |||
XM_017004507.1 | XP_016859996.1 | 34 | 3881 | |||
XM_017004508.1 | XP_016859997.1 | 34 | 3836 | |||
XM_017004509.1 | XP_016859998.1 | 19 | 2127 | |||
XM_017004510.1 | XP_016859999.1 | 15 | 2506 | |||
GenBank mRNA | AF227905.1 | - | 33 | 3682 | ||
AK023671.1 | - | 14 | 1518 | |||
AK074251.1 | - | 17 | 1846 | |||
AK074376.1 | - | 7 | 817 | |||
AK293066.1 | - | 33 | 3975 | |||
BC041098.1 | - | 33 | 3972 | 5492334 |