Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020135.2 | NP_064520.2 | 6 | 1885 | ||
NM_130395.2 | NP_569079.1 | 6 | 1810 | |||
GenBank mRNA | AB209723.1 | - | 6 | 1638 | ||
AF218313.1 | - | 6 | 1272 | |||
AK026179.1 | - | 5 | 790 | |||
AK223593.1 | - | 6 | 1819 | |||
AK315047.1 | - | 6 | 1977 | |||
AK315471.1 | - | 6 | 1779 | |||
BC018923.2 | - | 6 | 1771 | 4109718 | ||
U56249.1 | - | 2 | 130 |