Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020139.3 | NP_064524.3 | 5 | 402 | ||
XM_005263140.3 | XP_005263197.1 | 5 | 397 | |||
XM_006714274.3 | XP_006714337.1 | 5 | 397 | |||
XM_011532127.2 | XP_011530429.1 | 5 | 439 | |||
XM_011532128.2 | XP_011530430.1 | 5 | 439 | |||
XM_017008461.1 | XP_016863950.1 | 5 | 436 | |||
XM_017008462.1 | XP_016863951.1 | 5 | 436 | |||
GenBank mRNA | AF164790.1 | - | 5 | 366 | ||
AK023323.1 | - | 5 | 357 | |||
AK128463.1 | - | 6 | 1100 | |||
AK290160.1 | - | 5 | 358 | |||
AK291335.1 | - | 5 | 364 | |||
AK300626.1 | - | 5 | 247 | |||
AY358841.1 | - | 5 | 368 | |||
BC001953.1 | - | 5 | 395 | 2905279 | ||
BC037277.2 | - | 5 | 364 | 4838679 | ||
BC095414.1 | - | 5 | 370 | 5284436 | ||
BI544872.1 | - | 5 | 423 | |||
CR457309.1 | - | 4 | 297 |