Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020188.3 | NP_064573.1 | 3, 4 | 348 | ||
XM_017023461.1 | XP_016878950.1 | 4, 5 | 536 | |||
XM_017023467.1 | XP_016878956.1 | 5, 6 | 431 | |||
XM_017023468.1 | XP_016878957.1 | 4, 5 | 341 | |||
XM_017023469.1 | XP_016878958.1 | 4, 5 | 415 | |||
XM_017023470.1 | XP_016878959.1 | 4, 5 | 339 | |||
GenBank mRNA | AF201935.1 | - | 3, 4 | 324 | ||
AK123193.1 | - | 2, 3 | 1703 | |||
BC032631.1 | - | 3, 4 | 352 | 5399193 |