Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SDR39U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020195.2 | NP_064580.2 | 4 | 348 | ||
XM_011536975.1 | XP_011535277.1 | 4 | 392 | |||
GenBank mRNA | AF226050.1 | - | 3 | 622 | ||
AK123513.1 | - | 2 | 790 | |||
BC000989.2 | - | 4 | 348 | 2900140 | ||
BC008566.1 | - | 3 | 520 | |||
BI256676.1 | - | 2 | 211 | |||
DT220226.1 | - | 4 | 352 |