Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282786.1 | NP_001269715.1 | 3 | 299 | ||
NM_020200.6 | NP_064585.1 | 3 | 299 | |||
GenBank mRNA | AB209792.1 | - | 3 | 249 | ||
AF226056.1 | - | 3 | 247 | |||
AK021950.1 | - | 3 | 259 | |||
AK222674.1 | - | 3 | 258 | |||
AK294130.1 | - | 3 | 258 | |||
AK304055.1 | - | 3 | 258 | |||
AK309614.1 | - | 3 | 288 | |||
BC008662.2 | - | 3 | 261 | 3871911 |