Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020202.4 | NP_064587.1 | 7 | 608 | ||
GenBank mRNA | AF260334.1 | - | 7 | 607 | ||
AF284574.1 | - | 7 | 555 | |||
AK225979.1 | - | 6 | 1086 | |||
AK295835.1 | - | 6 | 475 | |||
AK313704.1 | - | 7 | 550 | |||
BC020620.1 | - | 7 | 565 | 4775062 | ||
BC107890.1 | - | 7 | 596 | 6648910 | ||
FJ224326.1 | - | 7 | 555 |