Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166283.1 | NP_001159755.1 | 3 | 778 | ||
NM_001166286.1 | NP_001159758.1 | 2 | 638 | |||
NM_001166287.1 | NP_001159759.1 | 3 | 752 | |||
NM_001166288.1 | NP_001159760.1 | 3 | 636 | |||
NM_001166289.1 | NP_001159761.1 | 3 | 656 | |||
NM_020211.2 | NP_064596.2 | 3 | 805 | |||
GenBank mRNA | AK074910.1 | - | 3 | 795 | ||
AK074966.1 | - | 2 | 612 | |||
AK074980.1 | - | 3 | 572 | |||
AK125204.1 | - | 4 | 761 | |||
AK291365.1 | - | 2 | 435 | |||
AK299363.1 | - | 3 | 654 | |||
AK309814.1 | - | 4 | 1018 | |||
AL136826.1 | - | 3 | 619 | |||
AL390083.1 | - | 2 | 561 | |||
BC015886.1 | - | 2 | 629 | |||
BC140838.1 | - | 3 | 583 | 9021706 | ||
BC151132.1 | - | 3 | 583 | 9057032 |