Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020223.3 | NP_064608.2 | 2 | 928 | ||
XM_017012450.1 | XP_016867939.1 | 2 | 923 | |||
XM_017012451.1 | XP_016867940.1 | 2 | 923 | |||
XM_017012452.1 | XP_016867941.1 | 2 | 923 | |||
XM_017012453.1 | XP_016867942.1 | 2 | 958 | |||
XM_017012454.1 | XP_016867943.1 | 2 | 938 | |||
GenBank mRNA | AB545605.1 | - | 2 | 928 | ||
AF533706.1 | - | 2 | 928 | |||
BC040074.1 | - | 2 | 784 | 4942737 |