Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 3 | 531 | ||
NM_020234.5 | NP_064619.2 | 4 | 659 | |||
XM_011521815.1 | XP_011520117.1 | 3 | 580 | |||
XM_017022419.1 | XP_016877908.1 | 3 | 578 | |||
XM_017022420.1 | XP_016877909.1 | 4 | 1440 | |||
XM_017022421.1 | XP_016877910.1 | 4 | 671 | |||
XM_017022422.1 | XP_016877911.1 | 3 | 1312 | |||
XM_017022423.1 | XP_016877912.1 | 3 | 580 | |||
XM_017022424.1 | XP_016877913.1 | 4 | 651 | |||
XM_017022425.1 | XP_016877914.1 | 4 | 651 | |||
XM_017022426.1 | XP_016877915.1 | 3 | 1312 | |||
XM_017022427.1 | XP_016877916.1 | 4 | 596 | |||
XM_017022428.1 | XP_016877917.1 | 4 | 601 | |||
XM_017022429.1 | XP_016877918.1 | 4 | 1547 | |||
XM_017022430.1 | XP_016877919.1 | 4 | 750 | |||
XM_017022431.1 | XP_016877920.1 | 3 | 192 | |||
GenBank mRNA | AF168717.1 | - | 3 | 508 | ||
AK290184.1 | - | 3 | 500 | |||
BC032535.2 | - | 4 | 599 | 5527713 | ||
BC093073.1 | - | 4 | 589 | 30326731 |