Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBX SIRNA Assays ›
Gene Symbol:
BBX
Gene Name:
BBX, HMG-box containing
Gene Aliases:
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location:
Chr.3: 107522936 - 107811329 on Build GRCh38
UniGene ID:
Hs.124366
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32540
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142568.2
NP_001136040.1
16
2773
NM_001276286.1
NP_001263215.1
15
1764
NM_020235.6
NP_064620.2
15
2683
XM_005247642.3
XP_005247699.1
16
2689
XM_005247643.3
XP_005247700.1
16
2701
XM_005247644.3
XP_005247701.1
17
2864
XM_011512999.2
XP_011511301.1
16
2773
XM_011513000.1
XP_011511302.1
16
2901
XM_011513001.1
XP_011511303.1
16
2741
XM_011513002.2
XP_011511304.1
16
2695
XM_011513003.2
XP_011511305.1
15
2683
XM_011513004.2
XP_011511306.1
15
2811
XM_011513005.2
XP_011511307.1
15
2605
XM_011513006.2
XP_011511308.1
15
1764
XM_011513007.2
XP_011511309.1
15
1686
XM_011513010.2
XP_011511312.1
14
1596
XM_017006881.1
XP_016862370.1
18
2914
XM_017006882.1
XP_016862371.1
16
2774
XM_017006883.1
XP_016862372.1
14
1674
GenBank mRNA
AF168718.1
-
4
351
AF276948.1
-
13
2471
AF454941.1
-
15
2676
AF454942.1
-
13
2556
AK025944.1
-
5
934
AK026082.1
-
6
1105
AK130627.1
-
6
759
AK291757.1
-
16
2719
AK314434.1
-
16
2724
AL136769.1
-
13
2556
AL834307.1
-
5
993
BC012837.1
-
6
1415
BC131718.1
-
15
1657
40123389
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
bone development
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline