Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020198.2 | NP_064583.2 | 3, 4 | 700 | ||
XM_005257527.2 | XP_005257584.1 | 3, 4 | 568 | |||
GenBank mRNA | AF113221.1 | - | 1, 2 | 46 | ||
AF226054.1 | - | 3, 4 | 548 | |||
AK027786.1 | - | 3, 4 | 542 | |||
AK027844.1 | - | 3, 4 | 529 | |||
AK075387.1 | - | 4, 5 | 629 | |||
AK225813.1 | - | 3, 4 | 570 | |||
AK314328.1 | - | 3, 4 | 555 | |||
BC008905.2 | - | 3, 4 | 522 | 3688068 |