Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300895.1 | NP_001287824.1 | 1 | 168 | ||
NM_145200.3 | NP_660201.1 | 1 | 168 | |||
XM_005274114.3 | XP_005274171.2 | 2 | 173 | |||
XM_011545181.2 | XP_011543483.1 | 2 | 173 | |||
XM_011545182.2 | XP_011543484.1 | 2 | 173 | |||
XM_017018025.1 | XP_016873514.1 | 2 | 376 | |||
GenBank mRNA | AL712439.1 | - | 1 | 24 | ||
AY039217.1 | - | 1 | 168 |