Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300743.1 | NP_001287672.1 | 11 | 1220 | ||
NM_020314.5 | NP_064710.4 | 11 | 1220 | |||
XM_005255435.3 | XP_005255492.1 | 11 | 1172 | |||
XM_006721065.3 | XP_006721128.1 | 11 | 1172 | |||
XM_011545896.2 | XP_011544198.1 | 10 | 850 | |||
XM_011545897.2 | XP_011544199.1 | 3 | 217 | |||
GenBank mRNA | AF461052.2 | - | 5 | 716 | ||
AK024693.1 | - | 7 | 490 | |||
AK095078.1 | - | 11 | 917 | |||
AK290286.1 | - | 11 | 917 | |||
AK294885.1 | - | 12 | 1275 | |||
AL832159.1 | - | 11 | 916 | |||
AL833428.1 | - | 11 | 909 | |||
BC014900.2 | - | 10 | 888 | 3909068 | ||
BC050464.1 | - | 11 | 960 | 6061139 | ||
BC058845.1 | - | 11 | 919 | 6452778 | ||
BX537867.1 | - | 11 | 932 |