Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020319.2 | NP_064715.1 | 2 | 334 | ||
XM_011515458.1 | XP_011513760.1 | 3 | 461 | |||
GenBank mRNA | AK001389.1 | - | 2 | 303 | ||
AK001740.1 | - | 2 | 272 | |||
AK023704.1 | - | 2 | 271 | |||
AL050390.1 | - | 2 | 275 | |||
BC035353.1 | - | 2 | 290 | 5185501 | ||
DA210027.1 | - | 2 | 335 |