Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 5 | 667 | ||
NM_001040195.1 | NP_001035285.1 | 4 | 632 | |||
NM_001040196.1 | NP_001035286.1 | 6 | 764 | |||
NM_001040197.1 | NP_001035287.1 | 6 | 785 | |||
NM_020350.4 | NP_065083.3 | 5 | 688 | |||
XM_011541799.2 | XP_011540101.2 | 6 | 681 | |||
XM_011541800.2 | XP_011540102.1 | 6 | 702 | |||
XM_011541802.1 | XP_011540104.1 | 7 | 778 | |||
GenBank mRNA | AF312374.1 | - | 5 | 619 | ||
AK290042.1 | - | 5 | 631 | |||
BC011946.1 | - | 5 | 573 | |||
BC017328.2 | - | 5 | 605 | 4546585 | ||
BQ672962.1 | - | 1 | 172 |